Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506151.2 | XP_003506199.1 | 2 - 3 | 558 | 90 | |
GenBank mRNA | JI886597.1 | - | 2 - 3 | 315 | 90 | |
JP043808.1 | - | 2 - 3 | 315 | 90 |