Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512368.2 | XP_003512416.1 | 2 - 3 | 318 | 62 | |
XM_007652860.1 | XP_007651050.1 | 2 - 3 | 319 | 62 | ||
GenBank mRNA | JI875984.1 | - | 2 - 3 | 253 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |