Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507990.2 | XP_003508038.1 | 4 - 5 | 645 | 100 | |
GenBank mRNA | JI867113.1 | - | 4 - 5 | 644 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |