Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3rf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503759.2 | XP_003503807.1 | 9 - 10 | 2163 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |