Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495118.1 | XP_003495166.1 | 8 - 9 | 1470 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |