Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502702.2 | XP_003502750.1 | 3 - 4 | 571 | 107 | |
GenBank mRNA | JI872736.1 | - | 3 - 4 | 574 | 107 | |
JP037977.1 | - | 3 - 4 | 574 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |