Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643065.1 | XP_007641255.1 | 4 - 5 | 834 | 89 | |
XM_007643066.1 | XP_007641256.1 | 4 - 5 | 692 | 89 | ||
GenBank mRNA | JP055791.1 | - | 3 - 4 | 630 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |