Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpine1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641676.1 | XP_007639866.1 | 7 - 8 | 1373 | 52 | |
GenBank mRNA | JI878505.1 | - | 7 - 8 | 1373 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |