Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511339.1 | XP_003511387.1 | 3 - 4 | 872 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |