Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512863.2 | XP_003512911.2 | 1 - 2 | 178 | 86 | |
GenBank mRNA | JI893058.1 | - | 1 - 2 | 178 | 86 | |
JP039980.1 | - | 1 - 2 | 178 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |