Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506875.2 | XP_003506923.1 | 4 - 5 | 792 | 61 | |
XM_003506876.2 | XP_003506924.1 | 4 - 5 | 795 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |