Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640808.1 | XP_007638998.1 | 3 - 4 | 1898 | 88 | |
GenBank mRNA | JP063271.1 | - | 2 - 3 | 330 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |