Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498087.2 | XP_003498135.1 | 4 - 5 | 846 | 82 | |
GenBank mRNA | JI871954.1 | - | 3 - 4 | 693 | 82 | |
JP041028.1 | - | 3 - 4 | 693 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |