Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007620746.1 | XP_007618936.1 | 3 - 4 | 162 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |