Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam86a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508119.2 | XP_003508167.1 | 5 - 6 | 491 | 105 | |
GenBank mRNA | JI888750.1 | - | 5 - 6 | 493 | 105 | |
JP050303.1 | - | 5 - 6 | 493 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |