Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007611028.1 | XP_007609218.1 | 1 - 2 | 152 | 62 | |
GenBank mRNA | JP041943.1 | - | 1 - 2 | 119 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |