Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tigit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497883.2 | XP_003497931.2 | 1 - 2 | 390 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |