Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100765863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511648.1 | XP_003511696.1 | 1 | 615 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |