Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647912.1 | XP_007646102.1 | 9 - 10 | 5927 | 70 | |
XM_007647913.1 | XP_007646103.1 | 9 - 10 | 5082 | 70 | ||
XM_007647914.1 | XP_007646104.1 | 9 - 10 | 4345 | 70 | ||
XM_007647915.1 | XP_007646105.1 | 9 - 10 | 4382 | 70 | ||
XM_007647916.1 | XP_007646106.1 | 8 - 9 | 6400 | 70 | ||
XM_007647917.1 | XP_007646107.1 | 9 - 10 | 4266 | 70 | ||
XM_007647918.1 | XP_007646108.1 | 10 - 11 | 2432 | 70 | ||
XM_007647919.1 | XP_007646109.1 | 9 - 10 | 2353 | 70 | ||
XM_007647920.1 | XP_007646110.1 | 8 - 9 | 2091 | 70 | ||
XM_007647921.1 | XP_007646111.1 | 8 - 9 | 1100 | 70 | ||
XM_007647922.1 | XP_007646112.1 | 8 - 9 | 1344 | 70 | ||
XM_007647923.1 | XP_007646113.1 | 7 - 8 | 6123 | 70 | ||
XM_007647924.1 | XP_007646114.1 | 7 - 8 | 1040 | 70 | ||
XM_007647925.1 | XP_007646115.1 | 7 - 8 | 940 | 70 | ||
XM_007647927.1 | XP_007646117.1 | 8 - 9 | 6183 | 70 | ||
GenBank mRNA | JI895875.1 | - | 3 - 4 | 512 | 70 | |
JP060073.1 | - | 3 - 4 | 512 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |