Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642752.1 | XP_007640942.1 | 1 - 2 | 273 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |