Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007626479.1 | XP_007624669.1 | 4 - 5 | 628 | 103 | |
GenBank mRNA | JI878707.1 | - | 3 - 4 | 418 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |