Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650487.1 | XP_007648677.1 | 10 - 11 | 1567 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |