Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650487.1 | XP_007648677.1 | 8 - 9 | 1284 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |