Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653644.1 | XP_007651834.1 | 3 - 4 | 594 | 96 | |
GenBank mRNA | JI868794.1 | - | 3 - 4 | 554 | 96 | |
JP043904.1 | - | 3 - 4 | 554 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |