Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652295.1 | XP_007650485.1 | 1 - 2 | 91 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |