Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100755558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502912.2 | XP_003502960.1 | 1 - 2 | 173 | 61 | |
XM_007643529.1 | XP_007641719.1 | 1 - 2 | 173 | 61 | ||
GenBank mRNA | JI893402.1 | - | 1 - 2 | 181 | 61 | |
JP043948.1 | - | 1 - 2 | 181 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |