Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650789.1 | XP_007648979.1 | 4 - 5 | 503 | 65 | |
GenBank mRNA | JI877430.1 | - | 1 - 2 | 138 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |