Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496443.2 | XP_003496491.2 | 7 - 8 | 1215 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |