Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsl1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496694.2 | XP_003496742.1 | 4 - 5 | 576 | 65 | |
GenBank mRNA | JI892047.1 | - | 4 - 5 | 561 | 65 | |
JP058574.1 | - | 4 - 5 | 576 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |