Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502655.2 | XP_003502703.1 | 14 - 15 | 2549 | 68 | |
XM_007643294.1 | XP_007641484.1 | 14 - 15 | 2354 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |