Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504160.2 | XP_003504208.1 | 1 - 2 | 169 | 52 | |
GenBank mRNA | JI885334.1 | - | 1 - 2 | 117 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |