Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643056.1 | XP_007641246.1 | 2 - 3 | 309 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |