Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653470.1 | XP_007651660.1 | 2 - 3 | 359 | 87 | |
GenBank mRNA | JP039565.1 | - | 2 - 3 | 359 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |