Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495765.2 | XP_003495813.2 | 2 | 119 | 102 | |
XM_007643900.1 | XP_007642090.1 | 2 | 117 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |