Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048388.1 | XP_017903877.1 | 4 - 5 | 647 | 110 | |
XM_018048389.1 | XP_017903878.1 | 4 - 5 | 553 | 110 | ||
XM_018048391.1 | XP_017903880.1 | 4 - 5 | 647 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |