Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681579.2 | XP_005681636.1 | 8 - 9 | 1441 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |