Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686914.3 | XP_005686971.1 | 1 - 2 | 369 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |