Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678474.3 | XP_005678531.2 | 4 - 5 | 475 | 68 | |
XM_013962787.2 | XP_013818241.1 | 4 - 5 | 476 | 68 | ||
XM_013962788.2 | XP_013818242.1 | 4 - 5 | 476 | 68 | ||
XM_018044891.1 | XP_017900380.1 | 4 - 5 | 476 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |