Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052833.1 | XP_017908322.1 | 3 - 4 | 681 | 87 | |
GenBank mRNA | JO649179.2 | - | 2 - 3 | 117 | 87 | |
JT660623.1 | - | 2 - 3 | 176 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |