Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062448.1 | XP_017917937.1 | 4 - 5 | 1429 | 96 | |
XM_018062449.1 | XP_017917938.1 | 4 - 5 | 1525 | 96 | ||
GenBank mRNA | JT746028.1 | - | 6 - 7 | 1378 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |