Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066939.1 | XP_017922428.1 | 7 - 8 | 1033 | 77 | |
GenBank mRNA | JO359473.2 | - | 1974 | 77 | ||
JO579201.1 | - | 2 - 3 | 3095 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |