Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056270.1 | XP_017911759.1 | 4 - 5 | 1996 | 76 | |
GenBank mRNA | JO498400.2 | - | 4 - 5 | 530 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |