Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966744.2 | XP_013822198.2 | 4 - 5 | 2239 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |