Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681180.2 | XP_005681237.2 | 5 - 6 | 1228 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |