Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684036.3 | XP_005684093.2 | 4 - 5 | 504 | 73 | |
XM_018052361.1 | XP_017907850.1 | 3 - 4 | 370 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |