Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041133.1 | XP_017896622.1 | 2 - 3 | 1026 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |