Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057498.1 | XP_017912987.1 | 3 - 4 | 1412 | 57 | |
XM_018057499.1 | XP_017912988.1 | 3 - 4 | 1488 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |