Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697186.3 | XP_005697243.3 | 1 - 2 | 790 | 73 | |
XM_018039656.1 | XP_017895145.1 | 1 - 2 | 791 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |