Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285739.1 | NP_001272668.1 | 5 - 6 | 900 | 84 | |
XM_018042656.1 | XP_017898145.1 | 6 - 7 | 980 | 84 | ||
XM_018042657.1 | XP_017898146.1 | 5 - 6 | 1329 | 84 | ||
GenBank mRNA | GQ918145.1 | - | 5 - 6 | 900 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |