Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285585.1 | NP_001272514.1 | 6 - 7 | 212 | 98 | |
XM_013964673.2 | XP_013820127.2 | 6 - 7 | 203 | 98 | ||
XM_013964674.2 | XP_013820128.2 | 6 - 7 | 262 | 98 | ||
XM_013964676.2 | XP_013820130.2 | 5 - 6 | 235 | 98 | ||
GenBank mRNA | AJ249995.1 | - | 5 - 6 | 182 | 98 | |
AJ289715.1 | - | 5 - 6 | 173 | 98 | ||
AJ289716.1 | - | 5 - 6 | 173 | 98 | ||
X65160.1 | - | 6 - 7 | 212 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |